Colony counters
Supplier: IUL Instruments SA
Jump to:
About this item
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
- Ø 100 or 120 mm biconvex lamp
- X2 magnification
220 V voltage, 50 Hz, 0,2 A.
2 Options Available Below
Product Details & Documents
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
- Ø 100 or 120 mm biconvex lamp
- X2 magnification
220 V voltage, 50 Hz, 0,2 A.
Product Family Options
Product Information
- AvailabilityPrice
- 710-0108Colony counter manual with magnifying glass ø 120 mm with lighting and supportColony counter manual with magnifying glass ø 120 mm with lighting and supportDiscontinued-
- 710-0106DOT colony counter with 100 mm magnifying glass without lightDOT colony counter with 100 mm magnifying glass without lightLog in for availability1 * 1 items1 048,00 €
- IULA90008700DOT colony counter with 120 mm magnifying glass with lightDOT colony counter with 120 mm magnifying glass with lightLog in for availability1 * 1 items1 251,00 €
Accessories
Product Information
- Catalog #AvailabilityPrice
- Centering Device 100 mm
Product Information
- Catalog #AvailabilityPrice
- Centering Device 80 mm
Product Information
- Catalog #AvailabilityPrice
- Centering Device 60 mm