Specifications
- Description:Colony counter, flash and go
- Seq:01
- Pk:1
Specifications
About this item
The first colony counter that grows and adapts to specific micro lab needs. It can be upgraded at any time to fit the level of automation, security and features required. The Flash & Go is the latest in a highly successful series of automatic colony counters. It combines sophisticated image processing and analysis with an innovative camera and light source to provide fast, accurate counts of colonies for a wide range of applications.
- It ensures excellent contrast between colonies and substrate
- For counting pour and spiral plates in 1 sec
- Every count under control: add or eliminate colonies with one click on the screen
- Delete an area by a simple draw with the mouse or with the touch screen pen
- Optimize detection with a single bar
- A high resolution CCD camera relays a live image of the sample to the computer screen
The 'Flash & Go' is ideal for laboratories engaged in drug development and production, food and milk testing, environmental monitoring and public health. Software prepared and recommended for Windows XP Tablet Edition. A specific software to measure the size of inhibitions zones or to cope with particles and to guarantee accurate counts, based on size sensitivity, shape and color can be added separately.