Portable balances, CS series
Supplier: OHAUS
Certificates
About this item
An economical balance with a rugged plastic housing and LCD display. Use via AC adapter or in battery-powered mode with power saving auto shut-off function and low battery indicator.
- Selection of weighing units
- Removable stainless steel platform
- Easy to use two button operation, external calibration
- Light and portable
- Mechanical and software overload protection
Delivery: Supplied with 3×AA batteries and AC adapter.
Frequently Bought Together