Orders LinkContactUsLinkComponent
Sterilisation autoclaves
Sterilisation autoclaves
Supplier:   SANOCLAV WOLF
Sterilisation autoclaves
Supplier:  SANOCLAV WOLF
Certificates
Sterilisation autoclaves
5 Options Available Below
Product Details & Documents
Autoclaves for sterilisation in tensioned steam and water.
  • Equipped with safety valve, overheat protection, drain valve and manometer
  • Pressure chamber with stainless steel lid and bowl
  • Microprocessor control
  • Filter system for exhaust air filtration
RS 232 interface with program for recording time and temperature on an external PC. Flexible PT 100 sensor for liquids (with the media sensor switched on, a pre- and post-heating temperature of 60 °C is maintained).
Safety interlock in La-MCS or La-VA-MCS, temperature-dependent at over 97 °C or locked at over 80 °C with the media sensor switched on.
Specifications for Product Family
Specifications
  • Catalog No:
  • 481-0264
  • 481-4640
  • 481-4639
  • SANC01032
  • SANC01033
  • Model:
  • LA-MCS-203
  • LA-MCS-204
  • LA-VA-MCS
  • LaM-3-20-MCS-J
  • LaM-4-20-MCS-J
  • Capacity:
  • 20 L
  • 20 L
  • 20 L
  • 20 L
  • 20 L
  • Max. pressure:
  • 3 bar
  • 4 bar
  • 4 bar
  • 3 bar
  • 4 bar
  • Max. temperature:
  • 143 °C
  • 151 °C
  • 151 °C
  • 143 °C
  • 151 °C
  • Int.Ø×H:
  • 260×400 mm
  • 260×400 mm
  • 260×400 mm
  • 260×400 mm
  • 260×400 mm
  • Ext.Ø×H:
  • 530×410 mm
  • 530×410 mm
  • W×D×H ext.:
  • 360×620×410 mm
  • 360×620×410 mm
  • 360×620×410 mm
  • Weight:
  • 27 kg
  • 27 kg
  • 27 kg
  • 20,5 kg
  • 20,5 kg
  • Heating power:
  • 1500 W
  • 1500 W
  • 1500 W
  • 1500 W
  • 1500 W
Specifications
Product Family Options
Product Information
  • Availability
    Price
  • 481-0264
    Autoclave, LA-MCS-203
    Autoclave, LA-MCS-203
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6 110,00 €
    6 110,00 €
    1 * 1 items
       6 110,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    481-0264
    Model
    LA-MCS-203
    Capacity
    20 L
    Max. pressure
    3 bar
    Max. temperature
    143 °C
    Int.Ø×H
    260×400 mm
    Ext.Ø×H
    W×D×H ext.
    360×620×410 mm
    Weight
    27 kg
    Heating power
    1500 W
  • 481-4640
    Autoclave, LA-MCS-204
    Autoclave, LA-MCS-204
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 6 220,00 €
    6 220,00 €
    1 * 1 items
       6 220,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    481-4640
    Model
    LA-MCS-204
    Capacity
    20 L
    Max. pressure
    4 bar
    Max. temperature
    151 °C
    Int.Ø×H
    260×400 mm
    Ext.Ø×H
    W×D×H ext.
    360×620×410 mm
    Weight
    27 kg
    Heating power
    1500 W
  • 481-4639
    Autoclave, LA-VA-MCS
    Autoclave, LA-VA-MCS
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7 040,00 €
    7 040,00 €
    1 * 1 items
       7 040,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    481-4639
    Model
    LA-VA-MCS
    Capacity
    20 L
    Max. pressure
    4 bar
    Max. temperature
    151 °C
    Int.Ø×H
    260×400 mm
    Ext.Ø×H
    W×D×H ext.
    360×620×410 mm
    Weight
    27 kg
    Heating power
    1500 W
  • SANC01032
    Autoclave, LaM-3-20-MCS-J
    Autoclave, LaM-3-20-MCS-J
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4 800,00 €
    4 800,00 €
    1 * 1 items
       4 800,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SANC01032
    Model
    LaM-3-20-MCS-J
    Capacity
    20 L
    Max. pressure
    3 bar
    Max. temperature
    143 °C
    Int.Ø×H
    260×400 mm
    Ext.Ø×H
    530×410 mm
    W×D×H ext.
    Weight
    20,5 kg
    Heating power
    1500 W
  • SANC01033
    Autoclave, LaM-4-20-MCS-J
    Autoclave, LaM-4-20-MCS-J
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4 960,00 €
    4 960,00 €
    1 * 1 items
       4 960,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SANC01033
    Model
    LaM-4-20-MCS-J
    Capacity
    20 L
    Max. pressure
    4 bar
    Max. temperature
    151 °C
    Int.Ø×H
    260×400 mm
    Ext.Ø×H
    530×410 mm
    W×D×H ext.
    Weight
    20,5 kg
    Heating power
    1500 W
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Flexible PT-100 floating sensor for liquids (sensor is switched on temperature of 60 °C is held)
    Flexible PT-100 floating sensor for liquids (sensor is switched on temperature of 60 °C is held)
    481-4632
    Log in for availability
    1 * 1 items - 505,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Safety lock (over 97 °C with solids and over 80 °C with liquids is the cover locked)
    Safety lock (over 97 °C with solids and over 80 °C with liquids is the cover locked)
    481-4633
    Log in for availability
    1 * 1 items - 1 500,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Interface (RS 232) with software to record the temperature and the time on a computer
    Interface (RS 232) with software to record the temperature and the time on a computer
    481-4634
    Log in for availability
    1 * 1 items - 533,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Bottom plate (A-30), aluminium eloxiert, with perforation and handle-hole, 230×30 mm
    Bottom plate (A-30), aluminium eloxiert, with perforation and handle-hole, 230×30 mm
    481-4512
    Log in for availability
    1 * 1 items - 54,70 €
Product Information
  • Catalog #
    Availability
    Price
  • Multi stage insert (T-5) 5-parts, 230×285 mm
    Multi stage insert (T-5) 5-parts, 230×285 mm
    SANC02005
    Log in for availability
    1 * 1 items - 186,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Wire basket (K 13), stainless steel, 230×170 mm
    Wire basket (K 13), stainless steel, 230×170 mm
    481-4513
    Log in for availability
    1 * 1 items - 203,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Wire basket (K 20), stainless steel, 230×320 mm
    Wire basket (K 20), stainless steel, 230×320 mm
    SANC02007
    Log in for availability
    1 * 1 items - 275,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Test tube rack (R 64) for 64 tubes, 175×175×180 mm
    Test tube rack (R 64) for 64 tubes, 175×175×180 mm
    SANC02008
    Log in for availability
    1 * 1 items - 207,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Extern exhaust bottle (KB)
    Extern exhaust bottle (KB)
    SANC02013
    Log in for availability
    1 * 1 items - 194,00 €