ContactUsLinkComponent
Wipe rolls, WYPALL* L40
Wipe rolls, WYPALL* L40
Catalog # 115-2051
Supplier:  KIMBERLY CLARK
undefined
Wipe rolls, WYPALL* L40
Catalog # 115-2051
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    1 roll of 750 sheets
  • Colour:
    White
  • Description:
    Large roll, 1 ply, perforated
  • L×W:
    315×340 mm
  • Seq:
    01
  • Pk:
    1 Roll
Specifications
About this item
These single ply wipes are made from AIRFLEX* material, a specialised fabric with a perfect balance of softness and strength, that is also thick and absorbent. Ideal for industrial tasks such as cleaning, process line wiping, soaking up large spills.
  • Made with double re-creped (DRC) base sheet technology
  • Large, perforated rolls for use in busy areas
  • Also suitable for use in healthcare for wiping patients’ hands and face
  • Can be used with a dispensing system to help control use and reduce wastage
Matching dispensers: 115-2038, 115-0045 and 115-1533.