Orders LinkContactUsLinkComponent
Shell/neckless vials, Chromacol™
Shell/neckless vials, Chromacol™
Supplier:   CHROMACOL
Shell/neckless vials, Chromacol™
Supplier:  CHROMACOL
Certificates
Shell/neckless vials, Chromacol™
Shell/neckless vials, Chromacol™
2 Options Available Below
Product Details & Documents
These vials are made from superiour quality 1st hydrolytic class glass (Type 1).
PE cap with starburst center design eases syringe needle penetration.
Product Family Options
With closures
Product Information
  • Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Cap material
    Availability
    Price
  • 548-0275
    Shell/neckless vial
    Shell/neckless vial
    4,00 ml
    Flat bottom
    15×46 mm
    Glass
    Clear
    Yes
    PE
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items
Without closures
Product Information
  • Capacity
    Base
    Ø×H
    Material
    Colour
    Cap included
    Cap material
    Availability
    Price
  • 548-1182
    Shell/neckless vial
    Shell/neckless vial
    1,00 ml
    Flat bottom
    8×40 mm
    Glass
    Clear
    No
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 163,00 €
    163,00 €
    1 * 500 items
       163,00 €