Orders LinkContactUsLinkComponent
Non-return valves
 :   KARTELL
Non-return valves
 :  KARTELL
Non-return valves
Non-return valves
PP, translucent. Prevent pressure loss and backflow. Maximum working pressure 2 bar and minimum working pressure  0,07 bar at room temperature (20 °C).
  • Excellent chemical resistance
  • Excellent thermal resistance, autoclavable
  • Tapered ends to ensure that tubes are held firmly
Product Family Options
  • Tubing size
  • 7,6 - 10 mm ID
    1 * 10 items
    1 * 10 items - 37,50 €
    37,50 €
    1 * 10 items
       37,50 €
  • 9,2 - 12,5 mm ID
    1 * 10 items
    1 * 10 items - 69,00 €
    69,00 €
    1 * 10 items
       69,00 €