ContactUsLinkComponent
Cutter for polymeric tubing, clean-cut
Cutter for polymeric tubing, clean-cut
Catalog # 554-1106
Supplier:  VICI JOUR RESEARCH
undefined
Cutter for polymeric tubing, clean-cut
Catalog # 554-1106
Supplier:  VICI JOUR RESEARCH
Specifications
  • Description:
    Cutter
  • Pk:
    1
Specifications
About this item
This cutter provides burr-free perpendicular cuts of tubing up to ¹/₈" in PEEK, PTFE, ETFE and other polymeric tubing.
  • Safety lock secures the blade
  • No distortion of outside or closing of inside of the tubing