Orders LinkContactUsLinkComponent
Sample tubes, disposable, Nunc™
Sample tubes, disposable, Nunc™
   Sustainable Options Available
Sample tubes, disposable, Nunc™
   Sustainable Options Available
Certificates
Sample tubes, disposable, Nunc™
6 Options Available Below
Product Details & Documents
PS, transparent.
  • Graduation marked
  • Good thermal resistance
  • Good chemical resistance
Tubes are suitable for a broad range of applications.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-0493
  • 391-8398
  • 391-8396
  • 391-8228
  • 525-0060
  • 391-8229
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
  • ISO 14001 (Certificate Registration No.CERT-IT000330)
Specifications
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Material
    Type
    Sterile
    Availability
    Price
  • 734-0493
    Round bottom, non sterile
    Round bottom, non sterile
    4 ml
    70 mm
    11 mm
    PS
    without stopper
    -
    Log in for availability
    Log in for availability
    1 * 3.600 items
    1 * 3.600 items - 288,00 €
    288,00 €
    1 * 3.600 items
       288,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    734-0493
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 391-8398
    Round bottom, non sterile
    Round bottom, non sterile
    5 ml
    75 mm
    12 mm
    PS
    without stopper
    -
    Log in for availability
    Log in for availability
    1 * 3.000 items
    1 * 3.000 items - 282,00 €
    282,00 €
    1 * 3.000 items
       282,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-8398
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 391-8396
    Round bottom, non sterile
    Round bottom, non sterile
    12 ml
    100 mm
    15 mm
    PS
    without stopper
    -
    Log in for availability
    Log in for availability
    1 * 1.200 items
    1 * 1.200 items - 154,00 €
    154,00 €
    1 * 1.200 items
       154,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-8396
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 391-8228
    Round bottom, sterile
    Round bottom, sterile
    12 ml
    100 mm
    15 mm
    PS
    with PE stopper
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 156,00 €
    156,00 €
    1 * 500 items
       156,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-8228
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 525-0060
    Conical bottom, non sterile
    Conical bottom, non sterile
    14 ml
    110 mm
    17 mm
    PS
    without stopper
    -
    Log in for availability
    Log in for availability
    1 * 1.800 items
    1 * 1.800 items - 375,00 €
    375,00 €
    1 * 1.800 items
       375,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0060
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
  • 391-8229
    Conical bottom, sterile
    Conical bottom, sterile
    14 ml
    110 mm
    17 mm
    PS
    with stopper
    +
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items - 171,00 €
    171,00 €
    1 * 500 items
       171,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    391-8229
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001 (Certificate Registration No.CERT-IT000330)
      Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Stoppers, PE, for tubes 12 mm external diameter
    Stoppers, PE, for tubes 12 mm external diameter
    732-2680
    Log in for availability
    1 * 3.000 items - 186,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Stoppers, PE, for tubes 15 mm external diameter
    Stoppers, PE, for tubes 15 mm external diameter
    391-8062
    Log in for availability
    1 * 2.400 items - 194,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Stoppers, PE, for tubes 11 mm external diameter
    Stoppers, PE, for tubes 11 mm external diameter
    391-8056
    Log in for availability
    1 * 3.600 items - 234,00 €