ContactUsLinkComponent
Centrifuge tubes, Nunc™
Centrifuge tubes, Nunc™
Catalog # 734-2186
   Low Manufacturing Impact
undefined
Centrifuge tubes, Nunc™
Catalog # 734-2186
   Low Manufacturing Impact
Specifications
  • Closure type:
    Screw cap
  • Ø×L:
    60×130 mm
  • Packed:
    4 per bag
  • Capacity:
    200 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    48
  • RCF max.:
    7000×g
Specifications
About this item
PP, conical bottom, graduated, sterile, autoclavable without cap.
  • Withstands forces up to 7000×g with adapter for 200 ml
  • Moulded graduations