Beaker tong, steel coated
231-4120
: BOCHEM
- Length:300 mm
- Pk:1
- Spanning area:32 - 140 mm
Steel, coated. For secure, easy handling of beakers and similar containers.
The jaws of the tongs are equipped with special silicone cushioning, which not only protect the containers from being damaged by the jaws, but also provide thermal insulation.