Orders LinkContactUsLinkComponent
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
BD Microlance™ 3 Needles
The BD Microlance™ 3 needles are designed for easy penetration, smooth gliding force during insertion/withdraw, and reduced cannula friction while sliding through tissue. These needles are available in a wide range of gauges, lengths, and bevel designs.
  • Recommended use: intradermal, subcutaneous, intramuscular, intravenous injections, and aspiration
  • BD Microlance™ needles can be connected to a luer slip adapter or luer-lock connector
  • Natural rubber latex is not part of the material formulation
  • Sterile, single-use design
Conventional needles with a regular size wall are less likely to flex than thin wall needles, due to the added cannula thickness. A thin wall needles inner diameter potentially allows use of a needle gauge that is one size smaller than that of a regular needle, while benefiting from the same flow rate.
 : Individually packed in blister film and medical grade paper.
Product Family Options
  • Description
    Gauge
    Length
    Ø
    Colour code
  • BD Microlance™ 3 needle, sterile
    G 27
    13 mm
    0,4 mm
    Grey
    1 * 100 items
    1 * 100 items - 7,80 €
    7,80 €
    1 * 100 items
       7,80 €
  • BD Microlance™ 3 needle, sterile
    G 30
    13 mm
    0,3 mm
    Yellow
    1 * 100 items
    1 * 100 items - 8,60 €
    8,60 €
    1 * 100 items
       8,60 €
  • BD Microlance™ 3 needle, thin wall, regular bevel
    G 30
    13 mm
    0,3 mm
    Yellow
  • BD Microlance™ 3 needle, sterile
    G 25
    16 mm
    0,5 mm
    Orange
    1 * 100 items
    1 * 100 items - 9,30 €
    9,30 €
    1 * 100 items
       9,30 €
  • BD Microlance™ 3 needle, sterile
    G 21
    16 mm
    0,8 mm
    Green
    1 * 100 items
    1 * 100 items - 9,70 €
    9,70 €
    1 * 100 items
       9,70 €
  • BD Microlance™ 3 needle, sterile
    G 25
    25 mm
    0,5 mm
    Orange
    1 * 100 items
    1 * 100 items - 9,30 €
    9,30 €
    1 * 100 items
       9,30 €
  • BD Microlance™ 3 needle, sterile
    G 23
    25 mm
    0,6 mm
    Blue
    1 * 100 items
    1 * 100 items - 8,90 €
    8,90 €
    1 * 100 items
       8,90 €
  • BD Microlance™ 3 needle, sterile
    G 21
    25 mm
    0,8 mm
    Green
    1 * 100 items
    1 * 100 items - 9,30 €
    9,30 €
    1 * 100 items
       9,30 €
  • BD Microlance™ 3 needle, sterile
    G 16
    40 mm
    1,6 mm
    White
    1 * 100 items
    1 * 100 items - 16,60 €
    16,60 €
    1 * 100 items
       16,60 €
  • BD Microlance™ 3 needle, thin wall, regular bevel
    G 22
    40 mm
    0,7 mm
    Black
  • BD Microlance™ 3 needle, sterile
    G 18
    40 mm
    Pink
    1 * 100 items
    1 * 100 items - 10,20 €
    10,20 €
    1 * 100 items
       10,20 €
  • BD Microlance™ 3 needle, sterile
    G 23
    40 mm
    0,64 mm
    Blue
    1 * 100 items
    1 * 100 items - 9,70 €
    9,70 €
    1 * 100 items
       9,70 €
  • BD Microlance™ 3 needle, sterile
    G 21
    40 mm
    0,8 mm
    Green
    1 * 100 items
    1 * 100 items - 9,70 €
    9,70 €
    1 * 100 items
       9,70 €
  • BD Microlance™ 3 needle, sterile
    G 21
    50 mm
    0,8 mm
    Green
    1 * 100 items
    1 * 100 items - 11,40 €
    11,40 €
    1 * 100 items
       11,40 €
  • BD Microlance™ 3 needle, sterile
    G 18
    50 mm
    1,20 mm
    Pink
    1 * 100 items
    1 * 100 items - 19,80 €
    19,80 €
    1 * 100 items
       19,80 €