Hypodermic needles, Neolus®
Supplier: TERUMO
Certificates
About this item
Sterile, disposable. Needles have been inspected meticulously and are of excellent quality.
- Normal, fine or ultra-fine wall, with 12° bevel points (except 613-4907 and 612-0130: 18,5°)
- Sterilised with ethylene oxide, comply with US Pharmacopeia requirements
- Standard Luer fittings, to AFNOR S 90 011 (6%)
Packaging: In individual sterile blister packs with colour coding.
Frequently Bought Together