Orders LinkContactUsLinkComponent
Stoppers with a conical ground joint
Transparent, 29/32
  217-0090
 :   KARTELL
Stoppers with a conical ground joint
  217-0090
 :  KARTELL
Stoppers with a conical ground joint
Stoppers with a conical ground joint
Transparent, 29/32
11,50 €
1 * 1 items
PP, transparent, with 2 hose attachments, 8 mm Ø. Ideal for ventilating or transferring air, gases or liquids in a container with a normal ground joint sleeve.
  • Sealing ring for excellent sealing
  • Autoclavable
  • Octogonal holding disc for easy loosening and removal of the stopper
  • NS:
    29/32
  • Colour:
    Transparent