Orders LinkContactUsLinkComponent
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
Supplier:   SciLabware
Stopcocks, Rotaflo®
Supplier:  SciLabware
Certificates
Stopcocks, Rotaflo®
15 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
Product Family Options
General purpose stopcocks, straight pattern
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 3 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 265,00 €
    265,00 €
    1 * 5 items
       265,00 €
  • 6 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 75,20 €
    75,20 €
    1 * 1 items
       75,20 €
  • 10 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 1 300,00 €
    1 300,00 €
    1 * 10 items
       1 300,00 €
General purpose stopcocks, right angle pattern
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 3 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 52,90 €
    52,90 €
    1 * 1 items
       52,90 €
  • 6 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 73,20 €
    73,20 €
    1 * 1 items
       73,20 €
High performance stopcocks, straight pattern
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 3 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 105,00 €
    105,00 €
    1 * 1 items
       105,00 €
  • 3 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 105,00 €
    105,00 €
    1 * 1 items
       105,00 €
  • 6 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 131,00 €
    131,00 €
    1 * 1 items
       131,00 €
  • 6 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 5 items
    1 * 5 items - 534,00 €
    534,00 €
    1 * 5 items
       534,00 €
  • 10 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 202,00 €
    202,00 €
    1 * 1 items
       202,00 €
High performance stopcocks, right angle pattern
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 3 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 108,00 €
    108,00 €
    1 * 1 items
       108,00 €
  • 3 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 108,00 €
    108,00 €
    1 * 1 items
       108,00 €
  • 6 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 136,00 €
    136,00 €
    1 * 1 items
       136,00 €
  • 6 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 107,00 €
    107,00 €
    1 * 1 items
       107,00 €
  • 10 mm
    13 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 188,00 €
    188,00 €
    1 * 1 items
       188,00 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 6 mm
    Replacement key, bore Ø 6 mm
    229-0230
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 2 items - 113,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 6 mm
    Replacement key, bore Ø 6 mm
    229-0231
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 156,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 6 mm
    Replacement locking ring for key with bore 6 mm
    BIBBLR6
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 37,20 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 10 mm
    Replacement key, bore Ø 10 mm
    229-0226
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 1 items - 72,30 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 3 mm
    Replacement key, bore Ø 3 mm
    229-0229
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 2 items - 80,60 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 3 mm
    Replacement locking ring for key with bore 3 mm
    BIBBLR3
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 25,40 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 10 mm
    Replacement key, bore Ø 10 mm
    229-0234
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 242,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 3 mm
    Replacement key, bore Ø 3 mm
    BIBBGP3K
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 99,40 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 10 mm
    Replacement locking ring for key with bore 10 mm
    BIBBLR10
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 35,40 €