Orders LinkContactUsLinkComponent
Chemist set
Chemist set
Set of 2 spatulas, 1 tweezers, 1 scissors
Catalog # 231-2177
Supplier:   RSG
Chemist set
Catalog # 231-2177
Supplier:  RSG
Chemist set
Chemist set
Set of 2 spatulas, 1 tweezers, 1 scissors
Price
55,00 €
Log in for availability
1 * 1 SET
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Useful combination of instruments used daily in the laboratory and in production.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Comes in practical imitation leather case
Each set includes a 130×4 mm micro double spatula, a 150×9 mm powder spatula, a pair of laboratory scissors (sharp/blunt, 130 mm long), as well as forceps (straight, sharp, 130 mm long).
Each set includes a 130×4 mm micro double spatula, a 150×9 mm powder spatula, a pair of laboratory scissors (sharp/blunt, 130 mm long), as well as forceps (straight, sharp, 130 mm long).
Specifications
  • Description:
    Set of 2 spatulas, 1 tweezers, 1 scissors