Orders LinkContactUsLinkComponent
Flat spatula, conical
Flat spatula, conical
Supplier:   NICKEL ELECTRO
Flat spatula, conical
Supplier:  NICKEL ELECTRO
Certificates
Flat spatula, conical
Flat spatula, conical
3 Options Available Below
Product Details & Documents
Stainless steel or pure nickel (99,6%). Multipurpose spatula for a wide range of uses.
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Smooth surfaces for fast, easy cleaning
Product Family Options
Product Information
  • Length
    Blade width
    Availability
    Price
  • 231-0127
    Stainless steel
    Stainless steel
    90 mm
    15/25 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,00 €
    7,00 €
    1 * 1 items
       7,00 €
  • NICK2122
    Stainless steel
    Stainless steel
    120 mm
    15/25 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 7,30 €
    7,30 €
    1 * 1 items
       7,30 €
  • Nickel
    180 mm
    20/35 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 71,60 €
    71,60 €
    1 * 1 items
       71,60 €