Orders LinkContactUsLinkComponent
Spoon spatulas
Spoon spatulas
Supplier:   KARTELL
Spoon spatulas
Supplier:  KARTELL
Certificates
Spoon spatulas
2 Options Available Below
Product Details & Documents
Polyamide, fibreglass reinforced, red.
  • Good chemical resistance
  • Heat resistance up to +100 °C (up to +180 °C for short periods)
  • Robust, flexible
Certifications: DIN 12890
Documents
Product Family Options
Product Information
  • Length
    Colour
    Availability
    Price
  • 180 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 42,40 €
    42,40 €
    1 * 10 items
       42,40 €
  • 210 mm
    Red
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 44,70 €
    44,70 €
    1 * 10 items
       44,70 €