Orders LinkContactUsLinkComponent
Ladle
Ladle
275 mm
Catalog # 231-1390
Supplier:   USBECK Laborgeräte
Ladle
Catalog # 231-1390
Ladle
Ladle
275 mm
Price
5,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel with wooden handle.
Ø 75 mm
Specifications
  • Length:
    275 mm