Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
Supplier:   Brand
Measuring scoops
Supplier:  Brand
Certificates
Measuring scoops
Measuring scoops
2 Options Available Below
Product Details & Documents
HDPE, natural coloured. With flat, bevelled end.
  • Good chemical resistance
  • Good thermal resistance
  • Robust, unbreakable construction
Product Family Options
Product Information
  • Capacity
    Length
    Availability
    Price
  • 500 ml
    350 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - 41,40 €
    41,40 €
    1 * 6 items
       41,40 €
  • 1000 ml
    400 mm
    Log in for availability
    Log in for availability
    1 * 6 items
    1 * 6 items - 58,20 €
    58,20 €
    1 * 6 items
       58,20 €