Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
Supplier:   HAMMACHER KARL
Laboratory scissors, straight
Supplier:  HAMMACHER KARL
Certificates
Laboratory scissors, straight
Laboratory scissors, straight
2 Options Available Below
Product Details & Documents
Stainless steel, with plastic handles.
  • Ideal for a wide range of laboratory applications
  • Corrosion resistant
  • Easy action
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Sharp/blunt
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 37,90 €
    37,90 €
    1 * 1 items
       37,90 €
  • Sharp/blunt
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 42,80 €
    42,80 €
    1 * 1 items
       42,80 €