Orders LinkContactUsLinkComponent
Scissors, multipurpose
Scissors, multipurpose
Supplier:   RSG
Scissors, multipurpose
Supplier:  RSG
Certificates
Scissors, multipurpose
Scissors, multipurpose
2 Options Available Below
Product Details & Documents
Stainless steel, with black plastic handles.
  • Corrosion resistant
  • Available in fine or normal design, with large handles
  • Easy action
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Laboratory, fine
    90 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 30,10 €
    30,10 €
    1 * 1 items
       30,10 €
  • Hobby, normal
    170 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 33,30 €
    33,30 €
    1 * 1 items
       33,30 €