Digital Solutions
Applications
Support
About
Slovenia
Orders Link
ContactUsLinkComponent
Scissors
Laboratory scissors
Laboratory scissors
With stopper lifter, sharp/blunt, 150 mm
233-2125
:
BOCHEM
Laboratory scissors
233-2125
:
BOCHEM
Laboratory scissors
With stopper lifter, sharp/blunt, 150 mm
9,00 €
1 * 1 items
-
+
Stainless steel.
Sharp-blunt
Corrosion-resistant
Autoclavable
With stopper lifter
Type:
With stopper lifter, sharp/blunt
Length:
150 mm
Acetone ≥99%, TECHNICAL
#: 20063.365
: VWR Chemicals