Orders LinkContactUsLinkComponent
Labels, control seals, close-it
Supplier:   BURKLE
Labels, control seals, close-it
Supplier:  BURKLE
Certificates
Labels, control seals, close-it
Labels, control seals, close-it
12 Options Available Below
Product Details & Documents
Designed to seal the holes in sacks or boxes created when samples are taken, the strong adhesive on the 'close-it' label allows adhesion to surfaces that are slightly moist, frozen or coated with powder. The aluminium backing creates a vapour barrier and seals the container hermetically.
  • Adheres extremely firmly
  • Can be written on
  • Colour coded for easy recognition
 : Rolls contain 500 (95×95 mm) or 250 (150×150 mm) stickers.
Product Family Options
95×95 mm
Product Information
  • Material
    Colour
    Dimensions
    Availability
    Price
  • 312-1154
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Black
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
  • 312-1151
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Red
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
  • 312-1153
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Yellow
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
  • 312-1152
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Green
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
  • 312-1155
    Control seals, blank
    Control seals, blank
    Aluminium/paper
    White
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
  • 312-1157
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Blue
    95×95 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 66,20 €
    66,20 €
    1 * 1 Roll
       66,20 €
150×150 mm
Product Information
  • Material
    Colour
    Dimensions
    Availability
    Price
  • 312-1161
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Red
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €
  • 312-1162
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Black
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €
  • 312-1163
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Yellow
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €
  • 312-1164
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Blue
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €
  • 312-1165
    Control seals, printed
    Control seals, printed
    Aluminium/paper
    Green
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €
  • 312-1166
    Control seals, blank
    Control seals, blank
    Aluminium/paper
    White
    150×150 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 92,70 €
    92,70 €
    1 * 1 Roll
       92,70 €