Orders LinkContactUsLinkComponent
VWR®, Securing key
VWR®, Securing key
Securing keys
Catalog # 241-0053
Supplier:   VWR Collection
VWR®, Securing key
Catalog # 241-0053
Supplier:  VWR Collection
VWR®, Securing key
VWR®, Securing key
Securing keys
Price
5,80 €
Log in for availability
1 * 5 items
Product Details & Documents
Toughened steel. This six-sided key is designed for use with Labomeca® bosses, joints and feet.
Hollow six-sided screws can be replaced with thumbscrews for assemblies which require frequent dismantling.
Specifications
  • Description:
    Securing keys