Orders LinkContactUsLinkComponent
Laboratory stands
Laboratory stands
Supplier:   KARTELL
Laboratory stands
Supplier:  KARTELL
Certificates
Laboratory stands
2 Options Available Below
Product Details & Documents
PP stand bases with rods made from chrome plated steel. Ideal for use as stands for burettes, funnels or separating funnels. Not recommended for heavier weights.
  • Rods are 250 mm long, 12 mm diameter and can be screwed together to reach the desired height
  • Model A: Stand base 300×150×15 mm with centre hole
  • Model B: Stand base 300×150×15 mm with off-centre hole
Ordering information: Each pack includes two rods measuring 250×12 mm. Extra rods can be ordered separately.
Product Family Options
Product Information
  • L×W
    Availability
    Price
  • 241-3215
    Model A
    Model A
    300×150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 126,00 €
    126,00 €
    1 * 1 items
       126,00 €
  • 241-3216
    Model B
    Model B
    300×150 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 127,00 €
    127,00 €
    1 * 1 items
       127,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Spare rod, chromium-plated steel, 12×250 mm
    Spare rod, chromium-plated steel, 12×250 mm
    241-3217
    Log in for availability
    1 * 1 items - 52,90 €