Orders LinkContactUsLinkComponent
Safety shields
Safety shields
Safety shields
Certificates
Safety shields
2 Options Available Below
Product Details & Documents
Protective shield made from acrylic glass to be used as splinter shield and splash guard.
  • Strong and stable design
  • Removable feet
Product Family Options
Product Information
  • Form
    W×D×H
    Availability
    Price
  • Straight
    630×8×550 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 328,00 €
    328,00 €
    1 * 1 items
       328,00 €
  • Straight
    900×10×670 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 465,00 €
    465,00 €
    1 * 1 items
       465,00 €