Orders LinkContactUsLinkComponent
Reagent sprayer
Reagent sprayer
Reagent sprayer, all glass, with 100 ml Erlenmeyer flask
Catalog # 552-0031
Supplier:   CAMAG
Reagent sprayer
Catalog # 552-0031
Supplier:  CAMAG
Reagent sprayer
Reagent sprayer
Reagent sprayer, all glass, with 100 ml Erlenmeyer flask
Price
217,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Glass atomiser. Cost efficient alternative for a TLC sprayer.
  • Can be used with compressed air or nitrogen
  • Reagents are atomised into a fine aerosol spray
Specifications
  • Description:
    Reagent sprayer, all glass, with 100 ml Erlenmeyer flask
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Rubber pump for 552-0031
    Rubber pump for 552-0031
    551-6103
    Log in for availability
    1 * 1 items - 30,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Erlenmeyer flask for 552-0031
    Erlenmeyer flask for 552-0031
    551-6102
    Log in for availability
    1 * 1 items - 38,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Spray head for 552-0031
    Spray head for 552-0031
    551-6101
    Log in for availability
    1 * 1 items - 113,00 €