Orders LinkContactUsLinkComponent
Pipette tips
   Sustainable Options Available
Supplier:   Sartorius
Pipette tips
Supplier:  Sartorius
   Sustainable Options Available
Certificates
Pipette tips
37 Options Available Below
Product Details & Documents
These pipette tips are suitable for many pipettes on the market.
  • Natural colour, premium grade PP, autoclavable
  • Excellent seal design for accurate, leak-tight sealing
  • Produced and packed in a dust-free environment under strictly maintained conditions
 : Available in bulk packaging, in autoclavable racks and in refills.
 : Article 613-4153 (BHIT780310) is designed for use with the Midi Plus Pipette Controller only.
Specifications for Product Family
Specifications
  • Catalog No:
  • 613-0678
  • 613-4161
  • 613-5099
  • 613-3024
  • 613-0679
  • 613-0915
  • 613-0916
  • 732-1556
  • 732-1557
  • 732-1558
  • 732-1559
  • BHITLH-X780200
  • BHITLH-X780201
  • 613-4140
  • 613-5100
  • 613-5101
  • 613-5102
  • 613-1665
  • 613-5106
  • 613-5107
  • 613-1791
  • 613-0687
  • 613-4172
  • 613-5105
  • 613-4175
  • 613-4176
  • 613-0680
  • 613-1792
  • 613-5109
  • 613-5111
  • 613-1793
  • 613-1794
  • 613-4149
  • 613-5090
  • 613-0677
  • 613-5089
  • BHITLH-799000
  • BHITLH-799004
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact|Product Transparency and Disclosure
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 140001; ACT Label
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
  • Renewable energy certificate; ISO 14001
Specifications
Product Family Options
Product Information
  • Tip style
    Volume
    Sterile
    Length
    Colour code
    Packed
    Availability
    Price
  • Standard
    0,1 - 10 µl
    -
    31,5 mm
    Grey
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 45,00 €
    45,00 €
    1 * 1.000 items
       45,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0678
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,1 - 10 µl
    -
    31,5 mm
    Grey
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 86,20 €
    86,20 €
    1 * 960 items
       86,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-4161
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,1 - 10 µl
    +
    31,5 mm
    Grey
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 122,00 €
    122,00 €
    1 * 960 items
       122,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5099
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,1 - 10 µl
    +
    31,5 mm
    Grey
    Refill pack, 20 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 1.920 items
    1 * 1.920 items - 182,00 €
    182,00 €
    1 * 1.920 items
       182,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-3024
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,1 - 10 µl
    -
    31,5 mm
    Grey
    Refill tower, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 74,50 €
    74,50 €
    1 * 960 items
       74,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0679
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    0,1 - 10 µl
    -
    46 mm
    Grey
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 87,10 €
    87,10 €
    1 * 960 items
       87,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0915
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    0,1 - 10 µl
    +
    46 mm
    Grey
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 120,00 €
    120,00 €
    1 * 960 items
       120,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0916
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,5 - 200 µl
    -
    51 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 79,30 €
    79,30 €
    1 * 960 items
       79,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1556
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,5 - 200 µl
    +
    51 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 96,80 €
    96,80 €
    1 * 960 items
       96,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1557
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,5 - 200 µl
    -
    51 mm
    Yellow
    Refill tower, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 67,60 €
    67,60 €
    1 * 960 items
       67,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1558
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    0,5 - 200 µl
    +
    51 mm
    Yellow
    Refill pack, 15 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 1.440 items
    1 * 1.440 items - 121,00 €
    121,00 €
    1 * 1.440 items
       121,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    732-1559
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended length
    0,5 - 200 µl
    -
    77,5 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 87,10 €
    87,10 €
    1 * 960 items
       87,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    BHITLH-X780200
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate
      Low Manufacturing Impact
  • Extended length
    0,5 - 200 µl
    +
    77,5 mm
    Yellow
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 104,00 €
    104,00 €
    1 * 960 items
       104,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    BHITLH-X780201
    Environmentally Preferable
    Environmental certification
  • Standard
    0,5 - 350 µl
    -
    54 mm
    Orange
    Bulk
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    613-4140
    Environmentally Preferable
    Environmental certification
  • Standard
    5 - 350 µl
    -
    54 mm
    Orange
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 83,40 €
    83,40 €
    1 * 960 items
       83,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5100
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    5 - 350 µl
    +
    54 mm
    Orange
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 96,80 €
    96,80 €
    1 * 960 items
       96,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5101
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    5 - 350 µl
    -
    54 mm
    Orange
    Refill tower, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 67,60 €
    67,60 €
    1 * 960 items
       67,60 €
    Specifications:
      Low Manufacturing Impact|Product Transparency and Disclosure
    More
    Specifications
    Cat. No.
    613-5102
    Environmentally Preferable
    Low Manufacturing Impact|Product Transparency and Disclosure
    Environmental certification
    Renewable energy certificate; ISO 140001; ACT Label
      Low Manufacturing Impact|Product Transparency and Disclosure
  • Standard
    5 - 350 µl
    +
    54 mm
    Orange
    Refill pack, 15 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 1.440 items
    1 * 1.440 items - 115,00 €
    115,00 €
    1 * 1.440 items
       115,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1665
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Wide bore
    10 - 1000 µl
    -
    68,5 mm
    Light blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 97,80 €
    97,80 €
    1 * 960 items
       97,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5106
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Wide bore
    10 - 1000 µl
    +
    68,5 mm
    Light blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 131,00 €
    131,00 €
    1 * 960 items
       131,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5107
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    10 - 1000 µl
    +
    71,5 mm
    Blue
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 80,00 €
    80,00 €
    1 * 960 items
       80,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1791
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    10 - 1000 µl
    -
    71,5 mm
    Blue
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 75,90 €
    75,90 €
    1 * 960 items
       75,90 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0687
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    10 - 1000 µl
    -
    71,5 mm
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 79,30 €
    79,30 €
    1 * 960 items
       79,30 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-4172
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    10 - 1000 µl
    +
    71,5 mm
    Blue
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 96,80 €
    96,80 €
    1 * 960 items
       96,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5105
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    50 - 1200 µl
    -
    71,5 mm
    Purple
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 98,70 €
    98,70 €
    1 * 960 items
       98,70 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-4175
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    50 - 1200 µl
    +
    71,5 mm
    Purple
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 116,00 €
    116,00 €
    1 * 960 items
       116,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-4176
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    50 - 1200 µl
    -
    71,5 mm
    Purple
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 79,20 €
    79,20 €
    1 * 960 items
       79,20 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0680
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    50 - 1200 µl
    +
    71,5 mm
    Purple
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 93,40 €
    93,40 €
    1 * 960 items
       93,40 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1792
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    50 - 1200 µl
    -
    90 mm
    Purple
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 104,00 €
    104,00 €
    1 * 960 items
       104,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5109
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    50 - 1200 µl
    +
    90 mm
    Purple
    10 racks, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 133,00 €
    133,00 €
    1 * 960 items
       133,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5111
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    50 - 1200 µl
    -
    90 mm
    Purple
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 88,60 €
    88,60 €
    1 * 960 items
       88,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1793
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Extended
    50 - 1200 µl
    +
    90 mm
    Purple
    Refill pack, 10 trays, 96 each
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 106,00 €
    106,00 €
    1 * 960 items
       106,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-1794
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    100 - 5000 µl
    -
    150 mm
    Green
    Bulk
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 19,80 €
    19,80 €
    1 * 100 items
       19,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-4149
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    100 - 5000 µl
    -
    150 mm
    Green
    Bulk
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 164,00 €
    164,00 €
    1 * 1.000 items
       164,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5090
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    100 - 5000 µl
    -
    150 mm
    Green
    Single tray (1×50)
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 16,80 €
    16,80 €
    1 * 50 items
       16,80 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-0677
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    100 - 5000 µl
    +
    150 mm
    Green
    Single tray (1×50)
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 27,10 €
    27,10 €
    1 * 50 items
       27,10 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    613-5089
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable energy certificate; ISO 14001
      Low Manufacturing Impact
  • Standard
    10000 µl
    -
    164 mm
    Red
    Single tray (4 x 35)
    Log in for availability
    Log in for availability
    1 * 140 items
    1 * 140 items - 129,00 €
    129,00 €
    1 * 140 items
       129,00 €
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    BHITLH-799000
    Environmentally Preferable
    Environmental certification
    New Product
  • Standard
    10000 µl
    -
    164 mm
    Red
    Bulk
    Log in for availability
    Log in for availability
    1 * 210 items
    1 * 210 items - 64,10 €
    64,10 €
    1 * 210 items
       64,10 €
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    BHITLH-799004
    Environmentally Preferable
    Environmental certification
    New Product