Orders LinkContactUsLinkComponent
Tip refill systems, Axygen®
Tip refill systems, Axygen®
Supplier:   Corning
Tip refill systems, Axygen®
Supplier:  Corning
Certificates
Tip refill systems, Axygen®
Tip refill systems, Axygen®
3 Options Available Below
Product Details & Documents
Transparent PP, autoclavable.
  • Easy to load refill racks
  • Certified RNase-, DNase- and endotoxin-free
  • Universal fit tips have a bevelled reference and graduated marks
Product Family Options
Gilson style pipette tips
Product Information
  • Volume
    Sterile
    Tip colour
    Packed
    Availability
    Price
  • 0,1 - 10 µl
    -
    Transparent
    10 racks, 96 each (stack rack)
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 65,50 €
    65,50 €
    1 * 960 items
       65,50 €
Universal fit pipette tips
Product Information
  • Volume
    Sterile
    Tip colour
    Packed
    Availability
    Price
  • 1 - 200 µl
    -
    Transparent
    10 racks, 96 each (stack rack)
    Log in for availability
    Log in for availability
    1 * 960 items
    1 * 960 items - 54,30 €
    54,30 €
    1 * 960 items
       54,30 €
  • 1 - 1200 µl
    -
    Transparent
    8 racks, 96 each (stack rack)
    Log in for availability
    Log in for availability
    1 * 768 items
    1 * 768 items - 73,90 €
    73,90 €
    1 * 768 items
       73,90 €
Empty refill boxes with lid
Product Information
  • Catalog #
    Description
    For use with
    Availability
    Price
  • Empty refill racks, RFL-R
    10 µl and 200 µl refill inserts
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 41,40 €
    41,40 €
    1 * 10 items
       41,40 €
Product Information
  • Catalog #
    Description
    For use with
    Availability
    Price
  • Empty refill racks, RFL-R-1200
    1200 µl refill inserts
    Log in for availability
    Log in for availability
    1 * 10 items
    1 * 10 items - 54,10 €
    54,10 €
    1 * 10 items
       54,10 €