Filter tips, sterile, ep Dualfilter T.I.P.S®
EPPE0030077.547RCH
- Colour code:Yellow
- Packed:5 racks, 96 each
- Sterile:+
- Volume:2 - 100 µl
- Length:53 mm
- Seq:07
- Tip style:PCR clean
- Pk:5 SET
Two-phase filter for ultimate aerosol protection: The white layer which faces the sample retains drops, splashes and aerosols; the blue layer above it forms a highly effective double-barrier, that reliably binds even biomolecules.
- Characterised by defined flow dynamics, low wettability and high thermal stability
- Certified pyrogen-free and PCR clean (free from human DNA, DNase, RNase and PCR-inhibiting substances)