Orders LinkContactUsLinkComponent
Slide trays
Slide trays
Supplier:   KARTELL
Slide trays
Supplier:  KARTELL
Certificates
Slide trays
2 Options Available Below
Product Details & Documents
Identification of different dyes on the slides.
  • Easy removal of slides
  • Slide trays for 76×26 mm slides
Colour: White
Product Family Options
Product Information
  • Colour
    Material
    W×D
    No. of slides
    Availability
    Price
  • 631-9004
    Slide tray
    Slide tray
    White
    PVC
    340×190 mm
    20
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 18,80 €
    18,80 €
    1 * 1 items
       18,80 €
  • 631-9144
    Slide tray
    Slide tray
    White
    PVC
    680×190 mm
    40
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 34,10 €
    34,10 €
    1 * 1 items
       34,10 €