Orders LinkContactUsLinkComponent
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, Nunc™
Microscope slides, Nunc™
These microscope slides are designed for applications in cell culture.
The surface treatment ensures optimal conditions for cell attachment and growth.
  • Catalog No:
  • 734-2070
  • 734-2071
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Product Family Options
  • Description
    L×W
  • Microscope slides, sterile, PS
    25×75 mm
    1 * 100 items
    1 * 100 items - 664,00 €
    664,00 €
    1 * 100 items
       664,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2070
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • Microscope slides, sterile, Permanox™
    27×75 mm
    1 * 100 items
    1 * 100 items - 553,00 €
    553,00 €
    1 * 100 items
       553,00 €
      Low Manufacturing Impact
    Specifications
    Cat. No.
    734-2071
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact