Orders LinkContactUsLinkComponent
Microscope slides
Microscope slides
Microscope slides
Supplier:  MARIENFELD
Certificates
Microscope slides
Microscope slides
5 Options Available Below
Product Details & Documents
Slides designed for in vitro diagnostic (IVD) applications.
  • Pre-washed and ready to use
  • Either ground or cut versions
  • Soda-lime glass, Hydrolytic Class 3
Extra-white slides are used for low auto-fluorescence applications.
Documents
Certificates
Search by Lot Number
Product Family Options
White
Product Information
  • Colour
    Thickness
    L×W
    Availability
    Price
  • 631-9460
    Plain slides, cut edges, soda-lime glass
    Plain slides, cut edges, soda-lime glass
    White
    1 mm
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 4,00 €
    4,00 €
    1 * 50 items
       4,00 €
  • 631-9461
    Double-frosted slides, cut edges, soda-lime glass
    Double-frosted slides, cut edges, soda-lime glass
    White
    1 mm
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 4,40 €
    4,40 €
    1 * 50 items
       4,40 €
  • 631-9462
    Plain slides, ground edges 90°, soda-lime glass
    Plain slides, ground edges 90°, soda-lime glass
    White
    1 mm
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 5,20 €
    5,20 €
    1 * 50 items
       5,20 €
  • 631-0015
    Double-frosted slides ground 90°, soda-lime glass
    Double-frosted slides ground 90°, soda-lime glass
    White
    1 mm
    76×26 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 6,70 €
    6,70 €
    1 * 50 items
       6,70 €
  • MARI1120250
    Slides, cut edges, soda-lime glass
    Slides, cut edges, soda-lime glass
    White
    1 mm
    75×75 mm
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 882,71 €
    882,71 €
    1 * 1.000 items
       882,71 €