ContactUsLinkComponent
Adhesion slides, Menzel Gläser, SuperFrost® Plus Gold
Adhesion slides, Menzel Gläser, SuperFrost® Plus Gold
Catalog # 630-1324
Supplier:  Epredia
undefined
Adhesion slides, Menzel Gläser, SuperFrost® Plus Gold
Catalog # 630-1324
Supplier:  Epredia
Specifications
  • Description:
    Adhesion slides, SuperFrost® Plus Gold
  • Pk:
    72
Specifications
About this item
Glass adhesive technology, which first attracts, and then chemically bonds fresh or formalin-fixed frozen tissue sections firmly to the surface.
  • Ground edges (90°) and frosted end for labelling
  • Attaches difficult to bond tissue sections
  • Washed and polished
Suitable for special stains, immunocytochemicals and in situ DNA hybridisation techniques.