ContactUsLinkComponent
Storage plates, 96-well, V-bottomed
  732-4942
   Low Manufacturing Impact
undefined
Storage plates, 96-well, V-bottomed
  732-4942
   Low Manufacturing Impact
  • Description:
    V-bottomed 96-well storage plate
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    100
PP.
  • V-bottomed well design for optimal sample recovery
  • Reinforced skirt for robotic manipulation
  • 0,2 ml maximum well volume