Gas lighter
Supplier: USBECK Laborgeräte
Certificates
About this item
Length: 155 mm.
Frequently Bought Together
Deep well and assay plates, Axygen®
Supplier: Corning
Cartridge burner, Teclu
Supplier: USBECK Laborgeräte
Digital burettes, Titrette®
Supplier: Brand
Bunsen burner, cartridge type
Supplier: USBECK Laborgeräte
Orthophosphoric acid 85% ≥85%, TECHNICAL
Supplier: VWR Chemicals
Repetitive pipette, Multipette® E3 / E3x
Supplier: EPPENDORF
VWR® Dropping Funnels, Cylincrical, with Pressure Balance
Supplier: VWR Collection
Thioacetamide 4% in aqueous solution Reag. Ph. Eur. 1089602
Supplier: VWR Chemicals
Cartridge burner
Supplier: USBECK Laborgeräte
EPA vials with PP cap
Supplier: EP SCIENTIFIC
Filter pump
Supplier: USBECK Laborgeräte
Eyewash solution
Supplier: Plum Safety
Glass ceramic laboratory protection plates, DURAN®
Supplier: DWK Life Sciences
Fuego SCS basic
Supplier: WARTEWIG
2-Propanol 70% ≥70% (v/v), GPR RECTAPUR®
Supplier: VWR Chemicals
Gas lighter
Supplier: USBECK Laborgeräte
Accessories
Supplier: Brand
ATTO 488 maleimide
Supplier: AAT BIOQUEST