Goggles, Elite
Anti-UV, anti-fog, Clear acetate, Transparent
Catalog # 111-0302
Supplier: Bollé Safety
Jump to:
Product Details & Documents
Comfortable goggles which can be worn with prescription eyewear.
- Extra-wide panoramic vision
- Indirect ventilation
- Pivoting, adjustable headband
Weight: 121 g
EN 166, EN 170
Certifications: EN 166, EN 170
Specifications
- Type:Anti-UV, anti-fog
- Colour:Transparent
- Lenses:Clear acetate