Heat resistant gloves, Topfire Kermel®
Catalog # 112-1077
Supplier: HONEYWELL SAFETY
Specifications
- Glove size:10
- Ambidextrous / hand-specific:Ambidextrous
- Disposable / reusable:Reusable
- Colour:Green/grey
- Length:370 mm
- Seq:02
- Thickness:1,1 - 1,5 mm
- EN norm:EN 388:2016 (3X43E), EN 407 (4443XX)
- Liner material:Canvas
- Pk:1 PAIR
Specifications
About this item
Gloves made from boucle Kermel® (1300 g/m²) with Canvas lining.
- Canvas-lined palm and back of hand
- Thumb stitching reinforced with Kevlar® fleece
- Split leather cuffs, length 130 mm
- Heat resistant up to 500 °C
Ideal for use in foundries and glassmaking industries.
Certifications: EN 388:2016 (3X43E), EN 407 (4443XX), Cat. III