Cleanroom gloves, polychloroprene, DermaShield® 73-711 / 73-721
: Ansell
Hand-specific, ergonomically designed gloves with a beaded cuff. Ideal for use in laboratories, pharmaceutical, biotech and medical device manufacturing.
- Sterile or non sterile, powder-free, bisque fingertips
- Optimal combination of product and worker protection
- Accelerator free: No allergies
- Softness and flexibility provides greater comfort compared to natural rubber latex
- High resistance to puncture and a wide range of chemicals
- Specific external surface treatment and constant monitoring of grip values
The sterile gloves are suitable for use in Class 100, ISO 5 or M3.5 cleanrooms. The non sterile gloves should be used in controlled but non sterile environments.
Length: 310 mm
Thickness: 0,18 mm (cuff), 0,175 mm (palm), 0,85 mm (fingertip)
Colour: Green
: 73-711: EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421; Cat. III; AQL 0,65
73-721: EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421; Cat. III; AQL 0,65
73-721: EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421; Cat. III; AQL 0,65
: The sterile gloves are packaged as single pairs in a poly wallet, sealed in a poly envelope; 10 pairs per sealed polybag; 5 sealed polybags to a master bag; 4 master bags per case liner; 200 pairs per carton.
The non sterile gloves are bulk packed: 20 right and left hand gloves are packed in separate compartments in a polybag; 1 polybag per vacuum-sealed polybag; 2 polybags per vacuum-sealed master polybag; 5 master polybags per case liner; 200 pairs per carton.
The non sterile gloves are bulk packed: 20 right and left hand gloves are packed in separate compartments in a polybag; 1 polybag per vacuum-sealed polybag; 2 polybags per vacuum-sealed master polybag; 5 master polybags per case liner; 200 pairs per carton.
- Catalog No:
- 112-3790
- 112-3789
- 112-3788
- 112-3792
- 112-3791
- 112-3804
- 112-3805
- 112-3806
- 112-3800
- 112-3801
- 112-3802
- 112-3803
- 112-3793
- 112-3794
- Model:
- Sterile (73-711)
- Sterile (73-711)
- Sterile (73-711)
- Sterile (73-711)
- Sterile (73-711)
- Non sterile (73-721)
- Non sterile (73-721)
- Non sterile (73-721)
- Non sterile (73-721)
- Non sterile (73-721)
- Non sterile (73-721)
- Non sterile (73-721)
- Sterile (73-711)
- Sterile (73-711)
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
- EN 455 (1/2), EN ISO 374-1:2016 Type A (KLMNPT), EN ISO 374-5:2016 VIRUS, EN 421
Frequently Bought Together