PE gloves
Gloves packed in poly bag. Suitable for use in non hazardous environments.
- Powder-free
- Textured for a reliable grip
- Ambidextrous
- No toxic admixtures, no heavy metal, free from formaldehyde
Colour: Transparent
- Catalog No:
- 112-0060
- 112-0061
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
Frequently Bought Together