Cleanroom gloves, nitrile, Kimtech™ G3
Supplier: KIMBERLY CLARK
Certificates
About this item
Ambidextrous gloves with beaded cuffs. Contain no natural rubber latex. Ideal for handling delicate equipment in microelectronics, semiconductor, optics and pharmaceutical industries.
- Non sterile, powder-free
- Textured fingertips to facilitate control and grip
- Beaded cuffs for added strength
- Static dissipative in use
- Silicone-free material
- Virus protection according to ISO 16604
Suitable for use in Class 1, ISO 3, M1.5 or higher cleanrooms.
Length: 300 mm
Thickness: 0,13 mm (palm)
Colour: White
Certifications: PPE Cat III (EU) 2016/425, EN 374-1 Type C (K), EN 374-5 (VIRUS), AQL 1.5.
Packaging: Packaged in cleanroom-compatible polyethylene. 100 gloves per double poly bag. 10 bags per box.
Related Information
Kimberly-Clark Cleanroom Gloves
Product Literature
KIMBHC61011_TDS_EN KIMBHC61012_TDS_EN
Datasheet
Frequently Bought Together