Vacuum filtration units
Supplier: Sartorius
Certificates
About this item
Borosilicate glass, aluminium clamp, autoclavable.
Vacuum filtration unit for clarification, particle testing and hybridisation tests.
516-7015: With glass frit filter support
511-0132: With PTFE-coated stainless steel screen filter support
Frequently Bought Together