Orders LinkContactUsLinkComponent
Funnel, disposable
Funnel, disposable
Disposable funnel, 60 mm
Catalog # 221-0083
Supplier:   MARYLAND PLASTICS
Funnel, disposable
Catalog # 221-0083
Supplier:  MARYLAND PLASTICS
Funnel, disposable
Funnel, disposable
Disposable funnel, 60 mm
Price
Inquire For Price
Log in for availability
1 * 100 items
Product Details & Documents
Transparent PP, single use
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Specifications
  • Funnel O-Ø:
    60 mm
  • Description:
    Disposable funnel