Orders LinkContactUsLinkComponent
Forceps
Forceps
Straight, blunt, Blue, 130 mm
Catalog # 232-0062
Supplier:   BOCHEM
Forceps
Catalog # 232-0062
Supplier:  BOCHEM
Forceps
Forceps
Straight, blunt, Blue, 130 mm
Price
0,80 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, blue. Universal forceps for a wide range of applications.
  • Autoclavable and sterilisable up to 180 °C
  • Serrated for improved grip
  • Cost-efficient
Specifications
  • Type:
    Straight, blunt
  • Colour:
    Blue
  • Length:
    130 mm