Orders LinkContactUsLinkComponent
Tissue forceps
Tissue forceps
Supplier:   BOCHEM
Tissue forceps
Supplier:  BOCHEM
Certificates
Tissue forceps
4 Options Available Below
Product Details & Documents
Stainless steel.
  • With small teeth (1:2) at the end of the blades
  • Designed for handling tissues with minimal trauma during surgery
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Tissue forceps 1:2
    145 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,60 €
    4,60 €
    1 * 1 items
       4,60 €
  • Tissue forceps 1:2
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,10 €
    4,10 €
    1 * 1 items
       4,10 €
  • Tissue forceps 1:2
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,30 €
    4,30 €
    1 * 1 items
       4,30 €
  • Tissue forceps 1:2
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 4,90 €
    4,90 €
    1 * 1 items
       4,90 €