Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
100 ml, 14/23, 0,2 ± ml
Catalog # SMIT129/B/100
Supplier:   SMITH SCIENTIFIC
Volumetric flasks, class B
Catalog # SMIT129/B/100
Supplier:  SMITH SCIENTIFIC
Volumetric flasks, class B
Volumetric flasks, class B
100 ml, 14/23, 0,2 ± ml
Price
45,20 €
Log in for availability
1 * 2 items
Product Details & Documents
Borosilicate glass, with plastic stopper.
Certifications: ISO 1042
Specifications
  • NS:
    14/23
  • Capacity:
    100 ml
  • Seq:
    10
  • Tolerance:
    0,2 ± ml