Orders LinkContactUsLinkComponent
Flasks, round bottom, with standard ground joint
Flasks, round bottom, with standard ground joint
Supplier:   VITLAB
Flasks, round bottom, with standard ground joint
Supplier:  VITLAB
Certificates
Flasks, round bottom, with standard ground joint
Flasks, round bottom, with standard ground joint
3 Options Available Below
Product Details & Documents
PFA, translucent.
  • Very good chemical resistance
  • Excellent thermal resistance from −270 to +250 °C
  • Autoclavable
Can also be used as safety flasks for rotary evaporators.
Product Family Options
Product Information
  • Capacity
    Socket size
    Height
    Ext.Ø
    Availability
    Price
  • 250 ml
    29/32 NS
    147 mm
    88 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 115,00 €
    115,00 €
    1 * 1 items
       115,00 €
  • 100 ml
    29/32 NS
    117 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 98,30 €
    98,30 €
    1 * 1 items
       98,30 €
  • 500 ml
    29/32 NS
    177 mm
    107 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 158,00 €
    158,00 €
    1 * 1 items
       158,00 €