Orders LinkContactUsLinkComponent
Erlenmeyer flasks with thread, DURAN®
Erlenmeyer flasks with thread, DURAN®
Supplier:  DWK Life Sciences
Certificates
Erlenmeyer flasks with thread, DURAN®
Erlenmeyer flasks with thread, DURAN®
4 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, without or with red PBT screw cap.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area. A PTFE-coated seal incorporated in the cap guarantees that the flasks are absolutely leakproof.
With printed Retrace Code (batch identification).
Product Family Options
With thread and screw cap
Product Information
  • Capacity
    Height
    Ext.Ø
    Thread
    Availability
    Price
  • 100 ml
    109 mm
    64 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 41,80 €
    41,80 €
    1 * 1 items
       41,80 €
  • 250 ml
    145 mm
    85 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 40,20 €
    40,20 €
    1 * 1 items
       40,20 €
  • 500 ml
    180 mm
    105 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 42,40 €
    42,40 €
    1 * 1 items
       42,40 €
  • 1000 ml
    225 mm
    131 mm
    GL 32
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 63,80 €
    63,80 €
    1 * 1 items
       63,80 €