Syringe filters, Acrodisc®, HT Tuffryn® membrane
Supplier: Cytiva (Formerly Pall Lab)
Certificates
About this item
Suitable for dilute biological fluids. Sterile versions are gamma irradiated and certified non pyrogenic. Low protein binding.
HT Tuffryn® polysulfone membrane; modified acrylic housing: 25 mm; PP housing: 13 mm and Acrodisc® PSF
Frequently Bought Together